Learn More
Fesoterodine-d7, TRC
Product Code. 30180395
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180395 25 mg 25mg
30180196 50 mg 50mg
30179772 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30180395 Supplier Toronto Research Chemicals Supplier No. TRCF32130325MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1126611-84-4
Molecular Formula C26H30D7NO3
Molecular Weight (g/mol) 418.62
SMILES [2H]C([2H])([2H])C([2H])(C(=O)Oc1ccc(CO)cc1[C@H](CCN(C(C)C)C(C)C)c2ccccc2)C([2H])([2H])[2H]
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C26H37NO3/c1-18(2)26(29)30-25-13-12-21(17-28)16-24(25)23(22-10-8-7-9-11-22)14-15-27(19(3)4)20(5)6/h7-13,16,18-20,23,28H,14-15,17H2,1-6H3/t23-/m1/s1/i1D3,2D3,18D
Formula Weight 418.3213
Chemical Name or Material Fesoterodine-d7
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.