Learn More
BINDER™ Access Port with Silicone Plug
BINDER™ Top Position Access Port is an accessory for use on the top of a variety of chambers and incubators.
Brand: BINDER™ 8012-1263
Additional Details : Weight : 0.00100kg
Description
- For use on top
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 10 mm, top | |
BINDER Chambers Series FD | |
10 mm |
Top | |
Silicone plug |