Learn More
BINDER™ Access Port with Silicone Plug
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series BD
BINDER Chambers Series KT
Diameter (Metric):
10 mm
50 mm
Unit Size:
Each
Description
- For use on top
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Specifications
| Description | Access port with silicone plug - 50 mm, top |
| Type | Access Port |
| For Use With (Equipment) | BINDER Chambers Series KT |
| Includes | Silicone Plug |
| Ports | Top Position |
| Diameter (English) | 2 in. |
| Diameter (Metric) | 50 mm |