missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech TOM1L1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 51017-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
TOM1L1 is an adapter protein involved in signaling pathways. Interacts with the SH2 and SH3 domains of various signaling proteins when it is phosphorylated. May promotes FYN activation, possibly by disrupting intramolecular SH3-dependent interactions.Specifications
TOM1L1 | |
Polyclonal | |
Unconjugated | |
TOM1L1 | |
OK/KNS CL.3, SRCASM, Target of Myb like protein 1, TOM1 like protein 1, TOM1L1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10040 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75674 | |
TOM1L1 | |
TOM1L1 Fusion Protein Ag0502 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |