missing translation for 'onlineSavingsMsg'
Learn More

TOM1L1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1457.58 NOK - 3943.39 NOK

Specifications

Antigen TOM1L1
Concentration 0.13 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16806285
View Documents
Proteintech
51017-1-AP-20UL
20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16896275
View Documents
Proteintech
51017-1-AP-150UL
150 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

TOM1L1 is an adapter protein involved in signaling pathways. Interacts with the SH2 and SH3 domains of various signaling proteins when it is phosphorylated. May promotes FYN activation, possibly by disrupting intramolecular SH3-dependent interactions.
Specifications

Specifications

TOM1L1
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human
O75674
10040
TOM1L1 Fusion Protein Ag0502
Primary
-20°C
TOM1L1
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
OK/KNS CL.3, SRCASM, Target of Myb like protein 1, TOM1 like protein 1, TOM1L1
TOM1L1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.