missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOM1L1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 51017-1-AP-20UL
This item is not returnable.
View return policy
Description
TOM1L1 is an adapter protein involved in signaling pathways. Interacts with the SH2 and SH3 domains of various signaling proteins when it is phosphorylated. May promotes FYN activation, possibly by disrupting intramolecular SH3-dependent interactions.Specifications
| TOM1L1 | |
| Polyclonal | |
| Unconjugated | |
| TOM1L1 | |
| OK/KNS CL.3, SRCASM, Target of Myb like protein 1, TOM1 like protein 1, TOM1L1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10040 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75674 | |
| TOM1L1 | |
| TOM1L1 Fusion Protein Ag0502 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction