missing translation for 'onlineSavingsMsg'
Learn More

TNFRSF4 Monoclonal Antibody (OTI7H9), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA812430S

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 17566864

  • 4065.15 NOK / 30µL
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TNFRSF4 Monoclonal antibody specifically detects TNFRSF4 in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications

Specifications

TNFRSF4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P43489
Mouse
Protein A/G
RUO
7293
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Western Blot
OTI7H9
Unconjugated
Tnfrsf4
ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l
Full length human recombinant protein of human TNFRSF4 (NP_003318) produced in HEK293T cell.
30 μL
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers