missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF4 Monoclonal Antibody (OTI7H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
2832.96 NOK - 6331.68 NOK
Specifications
| Antigen | TNFRSF4 |
|---|---|
| Clone | OTI7H9 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17566864
|
Origene Technologies
TA812430S |
30 μL |
2832.96 NOK
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17546864
|
Origene Technologies
TA812430 |
100 μL |
6331.68 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TNFRSF4 Monoclonal antibody specifically detects TNFRSF4 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| TNFRSF4 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l | |
| Full length human recombinant protein of human TNFRSF4 (NP_003318) produced in HEK293T cell. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Tnfrsf4 |
| OTI7H9 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P43489 | |
| 7293 | |
| IgG1 | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title