missing translation for 'onlineSavingsMsg'
Learn More

TMEM50B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15941-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16885344

  • 4593.76 NOK / 150µL
Estimated Shipment: 17-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TMEM50B gene ontology annotations related to this gene include late endosome to vacuole transport via multivesicular body sorting pathway.
Specifications

Specifications

TMEM50B
Polyclonal
Unconjugated
TMEM50B
C21orf4, HCVP7TP3, TMEM50B, transmembrane protein 50B
Rabbit
Antigen Affinity Chromatography
RUO
360698, 757, 77975
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P56557, Q9D1X9
Tmem50b
TMEM50B Fusion Protein Ag8220
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers