missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM50B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | TMEM50B |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16895344
|
Proteintech
15941-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16885344
|
Proteintech
15941-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TMEM50B gene ontology annotations related to this gene include late endosome to vacuole transport via multivesicular body sorting pathway.Specifications
TMEM50B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P56557, Q9D1X9 | |
360698, 757, 77975 | |
TMEM50B Fusion Protein Ag8220 | |
Primary | |
-20°C | |
TMEM50B |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C21orf4, HCVP7TP3, TMEM50B, transmembrane protein 50B | |
Tmem50b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |