missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM50B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15941-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
TMEM50B gene ontology annotations related to this gene include late endosome to vacuole transport via multivesicular body sorting pathway.Specifications
TMEM50B | |
Polyclonal | |
Unconjugated | |
TMEM50B | |
C21orf4, HCVP7TP3, TMEM50B, transmembrane protein 50B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
360698, 757, 77975 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P56557, Q9D1X9 | |
Tmem50b | |
TMEM50B Fusion Protein Ag8220 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |