missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tim23 Mouse anti-Human, Unconjugated, Clone: 3H10F5, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | Tim23 |
---|---|
Clone | 3H10F5 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16800226
|
Proteintech
67535-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16890216
|
Proteintech
67535-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Tim23 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Tim23. This antibody recognizes human, mouse, rat antigen. The Tim23 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Tim23 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
10431 | |
Primary |
3H10F5 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |