Learn More
Tim23 Mouse anti-Human, Unconjugated, Clone: 3H10F5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67535-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16890216
  • 4593.76 NOK / 150µL
Estimated Shipment: 27-05-2024
Description
The Tim23 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Tim23. This antibody recognizes human, mouse, rat antigen. The Tim23 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
Tim23
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence
3H10F5
Unconjugated
150 μL
10431
IgG1