Learn More
TIGIT Monoclonal Antibody (OTI8G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812978S
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| TIGIT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q495A1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 201633 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| OTI8G7 | |
| Unconjugated | |
| TIGIT | |
| ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM | |
| Full length human recombinant protein of human TIGIT (NP_776160) produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.