missing translation for 'onlineSavingsMsg'
Learn More

TIGIT Monoclonal Antibody (OTI8G7), TrueMAB™, OriGene

Product Code. p-7225523
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
Unit Size:
100µL
30µL
Product Code. Quantity unitSize
17577574 30 μL 30µL
17557574 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17577574

Brand: Origene Technologies TA812978S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.

TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen TIGIT
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI8G7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TIGIT
Gene Accession No. Q495A1
Gene Alias ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM
Host Species Mouse
Immunogen Full length human recombinant protein of human TIGIT (NP_776160) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201633
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.