Learn More
Novus Biologicals™ THSD1 Antibody Pair [HRP]
Product Code. 18600041 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Product Code. 18600041
Brand: Novus Biologicals™ NBP2795755Plates
Polyclonal/Monoclonal Antibody Pair
This solid phase sandwich ELISA utilizes an antibody specific for Mouse THSD1.
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 62.5 pg/mL
Assay Sensitivity 62.5-4000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Mouse THSD1.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias thrombospondin type-1 domain-containing protein 1, thrombospondin, type I, domain 1, thrombospondin, type I, domain containing 1,4833423O18Rik, TMTSPMGC74971, Transmembrane molecule with thrombospondin module, UNQ3010
Gene ID (Entrez) 55901
Gene Symbol THSD1
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.25 mg/mL rabbit anti-THSD1 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-protector, pH 7.4, store at 4°C).
  • Capture Antibody: 0.5 mg/mL of rabbit anti-THSD1 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 95 ng of recombinant THSD1.
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Cancer
    Storage Requirements Storage is content dependent.
    Target THSD1
    Technique 1. Capture Antibody: Dilute to a working concentration of 1 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 4 ng/mL is recommended.