missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THOC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55178-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The TREX multiprotein complex binds specifically to spliced mRNAs to facilitate mRNA export. The protein encoded by this gene is a member of the THO complex, a subset of the TREX complex. The encoded protein interacts with the THOC1 protein.Specifications
THOC2 | |
Polyclonal | |
Unconjugated | |
THOC2 | |
THOC2 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NI27 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57187 | |
-20°C | |
Liquid |