missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THOC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | THOC2 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Preis | Menge & Verfügbarkeit | |||||
16896655
|
Proteintech
55178-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16886655
|
Proteintech
55178-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
The TREX multiprotein complex binds specifically to spliced mRNAs to facilitate mRNA export. The protein encoded by this gene is a member of the THO complex, a subset of the TREX complex. The encoded protein interacts with the THOC1 protein.Spezifikation
THOC2 | |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
Q8NI27 | |
THOC2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
57187 | |
Peptide | |
Primary | |
-20°C | |
THOC2 |