Learn More
Fisherbrand™ Thermographic Measurements Temperature Strip
Product Code. 11592503 Shop All Fisherbrand Products
Encompass_Preferred
Click to view available options
Temperature Range (Metric):
121°C to 160°C
77°C to 116°C
Unit Size:
Pack of 10
Product Code. 11592503
This item requires a price update prior to being ordered.
Request a Price for this item here.
The standard 8 level range covers 40°C to 260°C for general purpose temperature monitoring.
Specifications
For Use With (Application) For general purpose temperature monitoring
Product Type Temperature Strip
Max. Temperature (Metric) 116°C
Min. Temperature (Metric) 77°C
Temperature Range (Metric) 77°C to 116°C
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.