Learn More
Unity Lab Services™ Thermo Scientific™ Ultra-Low Temperature Freezers Validation Service
Product Code. 15671599
Change view
Click to view available options
Service Offering:
IQ/OQ Validation
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Service Offering Unit Size
15671599 IQ/OQ Validation Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15671599 Supplier Unity Lab Services™ Supplier No. IOQPCKE89003810
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Ultra-Low Temprature Freezers Models: Thermo Scientific TSX-series (post 2018), Forma 89000-series, HeraFreeze HLE, Revco RLE, Thermo Scientific TLE
IQ
  • Unpacking, Assembly and Installation Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
  • Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ System Component Information Detailed configuration information for each system component have been recorded.
  • OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
  • Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment.
- Key Tests: Door Ajar, Power Failure, Temperature Alerts
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific ULT Freezers: TSX-series (post 2018), Forma 89000-series, HeraFreeze HLE, Revco RLE, TLE
Description Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification.
Service Offering IQ/OQ Validation
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service