Learn More
Thermo Scientific™ TSX Series High-Performance Lab Refrigerators
BIOPHARMA
Product Code. p-6960052 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (English):
11.5 cu. ft.
23 cu. ft.
29.2 cu. ft.
45.8 cu. ft.
51.1 cu. ft.
Door Style:
Double Glass
Double Solid
Glass
Sliding Glass
Solid
Plug Type:
CEE 7/7
NEMA 6-15
Electrical Requirements:
208-230V, 50/60Hz
208-230V, 50Hz
208-230V, 60Hz
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) Door Style Plug Type Electrical Requirements Unit Size
15993610 45.8 cu. ft. Sliding Glass CEE 7/7 208-230V, 50Hz Each
15913620 51.1 cu. ft. Double Glass CEE 7/7 208-230V, 50Hz Each
17269009 51.1 cu. ft. Double Solid NEMA 6-15 208-230V, 60Hz Each
15933620 51.1 cu. ft. Double Solid CEE 7/7 208-230V, 50Hz Each
16317181 11.5 cu. ft. Glass CEE 7/7 208-230V, 50Hz Each
16337181 11.5 cu. ft. Solid CEE 7/7 208-230V, 50Hz Each
16397181 29.2 cu. ft. Glass CEE 7/7 208-230V, 50Hz Each
16317191 29.2 cu. ft. Solid CEE 7/7 208-230V, 50Hz Each
15618416 23 cu. ft. Glass CEE 7/7 208-230V, 50/60Hz Each
15638416 23 cu. ft. Solid CEE 7/7 208-230V, 50/60Hz Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 15993610 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' TSX4505GV
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Innovative refrigeration solutions for healthcare and research needs.
Thermo Scientific™ TSX Series high-performance lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade storage requiring 3° to 7°C. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Featuring over 450W of reserve refrigeration capacity, the TSX series has the power to protect your product from temperature swings as a result of frequent and prolonged door access, large product loads, and other activities that introduce hear to the cabinet.
The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
The drive for sample integrity and security
  • Designed to safely store media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade products requiring a temperature range of 3°C to 7°C
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance
Enabling compliance
  • Tested to GMP Clean Room Class A / ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
  • Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863
Designed for an ergonomic and comfortable experience
  • Ergonomically designed door handle with a soft-touch grip
  • The controller is angled to help prevent neck strain
  • Convenient, easy to clean and adjustable epoxy coated full and half shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Bright, LED interior lighting
  • Self-closing multipaned glass or solid doors with 90° stop to assist with inventory loads
Verified sustainability
  • US EPA ENERGY STAR Certified with up to 40% less energy usage than conventional refrigerant models*
  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life. **
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility
Ready for connectivity and monitoring
  • Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always—and anywhere— know the safety of your samples and the performance of your refrigerator or freezer.
  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems
Make it your own
  • Alternate Door swing configuration and door types
  • Extra and Alternate wire baskets, shelves, and drawers
  • Free-standing or factory-installed chart recorders
  • Stainless Steel interior finish
  • E-Lock Adapter kits
TRUSTED_SUSTAINABILITY
Specifications
Type High-Performance Refrigerator
Capacity (English) 45.8 cu. ft.
Capacity (Metric) 1297 L
Temperature Range 3°C to 7°C
Temperature (Setpoint) 3°-7°C (5°C)
Refrigerant R290
Door Style Sliding Glass
No. of Doors 2
Shelves 8 Wire
Cabinet Material Cold-rolled Steel
Exterior Finish Painted Steel
Certifications/Compliance UL, cUL, CE
Style Upright
Display Microprocessor
Product Line TSX Series
Warranty Warranty varies by country, please contact us for details
Defrost Automatic
Monitoring Options Alarm, Recorder
Chart Recorder Optional
Smart View Compatibility Smart-Vue Compatible
Compressor V-drive
Dimensions (D x W x H) Interior 28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Dimensions (L x W x H) Exterior 36.6 x 56.5 x 78.5 in (93.1 x 143.5 x 199.4 cm)
Depth (English) Exterior 36.6 in.
Depth (English) Interior 28.5 in.
Depth (Metric) Exterior 93.1 cm
Depth (Metric) Interior 72.4 cm
Height (English) Exterior 78.5 in.
Height (English) Interior 58 in.
Height (Metric) Exterior 199.4 cm
Height (Metric) Interior 147.3 cm
Width (English) Exterior 56.5 in.
Width (English) Interior 52.5 in.
Width (Metric) Exterior 143.5 cm
Width (Metric) Interior 133.4 cm
Amperage 15 A (CB)
Plug Type CEE 7/7
Voltage 208/230 V
Electrical Requirements 208-230V, 50Hz
Frequency 50/60 Hz
Green Features ENERGY STAR
Shipping Weight (English) 611 lb.
Shipping Weight (Metric) 277 kg
Show More Show Less
*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.