Learn More
Fisherbrand™ accumet™ Chloride Combination Electrode - Mercury-Free
Product Code. 11500154 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Connector Types:
BNC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
11500154 BNC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11500154
  • ONLINE EXCLUSIVE PRICE
    11940.00 NOK
    8686.27 NOK / Pack of 1
    Save 3253.73 NOK
    27% Off
View all promotions
Estimated Shipment: 13-01-2027
Typical applications include water/wastewater, soil, dairy, meats, tomato/vegetable products.
Method ASTM/AOAC approved
TRUSTED_SUSTAINABILITY
Specifications
Applications Chloride
Connector Types BNC
Junction Sure-Flow Junction
Material Plastic
For Use With (Equipment) ISE Meter with a BNC Connection
Temperature (Metric) Operating 0°C to 80°C
Type Chloride Combination Electrode