Learn More
Unity Lab Services™ Thermo Scientific Centrifugation Validation Service with Cycle Testing for Superspeed and Ultra centrifuges: Thermo Scientific, Sorvall
Product Code. 15691539
Change view
Click to view available options
For Use With (Equipment):
Sorvall Superspeed and Ultra Centrifuges
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15691539 Sorvall Superspeed and Ultra Centrifuges Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15691539 Supplier Unity Lab Services™ Supplier No. IOPQPCKE89003493
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Thermo Scientific Centrifugation Validation Service with Cycle Testing for Superspeed and Ultra centrifuges: Thermo Scientific, Sorvall
IQ
  • Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
  • Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ
  • System Component Information Detailed configuration information for each system component have been recorded.
  • OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
  • Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment. - Key Tests: Min/Max Speed, Brief/Extended Time, Temperature for Refrigerated Models
Cycle Testing
  • Point-In-Time test exercise of pre-defined duration that surpasses singular calibration and routine speed, time, and temperature monitoring activities for ensuring that the equipment is performing in accordance with the intended use of the system.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Sorvall Superspeed and Ultra Centrifuges
Description Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform min/max speed, brief/extended time and temperature testing (for refrigerated models). Cycle Testing: Point-In-Time test exercise of pre-defined duration that surpasses singular calibration and routine speed, time, and temperature monitoring activities.
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service