Learn More
Thermographic Measurements Thermax 10-Level Temperature Indicator Strips, 132°C to 182°C (270°F to 360°F)
Product Code. 17729398
Change view
Click to view available options
Temperature Range (Metric):
132°C to 182°C
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
17729398 132°C to 182°C Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17729398 Supplier Thermographic Measurements Supplier No. M10STHERGCD01P1
  • ONLINE EXCLUSIVE PRICE
    159.00 NOK
    131.60 NOK / Pack of 10
    Save 27.40 NOK
    17% Off
View all promotions
Estimated Shipment: 25-11-2026
Single use, irreversible threshold indicator reports the highest temperature reached.
  • Immediate response
  • Permanent record of highest temperature achieved
  • Oil, water, and steam resistant
  • Inexpensive and easy to use
  • Self-adhesive
Specifications
Certifications/Compliance REACH Compliant, RoHS Compliant, ISO9001
Disposable Single-use
For Use With (Application) Surface temperature measurement
Height (English) 2.01 in.
Height (Metric) 51 mm
Includes Indicator Strip
Length (Metric) 51 mm
Material Plastic
Weight (Metric) 9 g
Width (English) 4.09 in.
Product Type Self-adhesive Temperature Indicator Strip
Length (English) 2.01 in.
Max. Temperature (English) 360°F
Max. Temperature (Metric) 182°C
Min. Temperature (English) 270°F
Min. Temperature (Metric) 132°C
Temperature Range (English) 270°F to 360°F
Temperature Range (Metric) 132°C to 182°C
Calibrated No
For Use With (Equipment) Surfaces
Accuracy ±1.5°C less than 154°C and ±4°C above
Subdivision 10 Levels
Display Vertical
Width (Metric) 104 mm
Scale Celcius and Farenheit