Learn More
Ebro Electronic™ Thermocouple Wire Probes
GREENER_CHOICE
Product Code. 10461331
Change view
Click to view available options
Length (Metric):
150 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10461331 150 mm Each
1 options
Product Code. 10461331
Brand: Ebro Electronic™ 13410646
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermal wire probes for measurement of air, oven and gas temperatures
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RoHS Directive 2011/65/EU
Disposable Single-use
For Use With (Application) Thermal wire probes, measurement of air, oven and gas temperatures.
Height (Metric) 20 mm
Immersion Type Total or Partial
Length (Metric) 150 mm
Material Metal
Model TPN 600
Probe Type NiCrNi
Resolution 0.1°C
Weight (Metric) 0.06 kg
Product Type Thermal Wire Probe
Max. Temperature (Metric) 400°C
Min. Temperature (Metric) -50°C
Temperature Range (Metric) -50°C to +400°C
For Use With (Equipment) TFN 520
Accuracy DIN IEC 584, Class 2
Width (Metric) 150 mm