missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Thalidomide-O-amido-PEG2-C2-NH2 TFA

Product Code. 16440146
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16440146 2 g 2g
16491606 500 mg 500mg
16471179 1 g 1g
16465496 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16440146 Supplier MedChemExpress Supplier No. HY112617A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Thalidomide-O-amido-PEG2-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 2-unit PEG linker used in PROTAC technology.

Specifications

Molecular Weight (g/mol) 576.48 g/mol
CAS 1957235-75-4
Content And Storage 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Shipping Condition Room temperature
Quantity 2 g
Synonym Cereblon Ligand-Linker Conjugates 10 TFA ∣ E3 Ligase Ligand-Linker Conjugates 24 TFA
Solubility H2O : 150 mg/mL (260.20 mM; Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.