missing translation for 'onlineSavingsMsg'
Learn More

TGFBI / BIGH3 Mouse anti-Human, Mouse, Clone: 3E11D11, Proteintech

Mouse Monoclonal Antibody

1710.33 NOK - 4836.10 NOK

Specifications

Antigen TGFBI / BIGH3
Clone 3E11D11
Concentration 0.39 mg/mL
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16846965
View Documents
Proteintech
60007-1-IG-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16836965
View Documents
Proteintech
60007-1-IG-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes an RGD-containing protein that binds to type I, II and IV collagens. The RGD motif is found in many extracellular matrix proteins modulating cell adhesion and serves as a ligand recognition sequence for several integrins. This protein plays a role in cell-collagen interactions and may be involved in endochondrial bone formation in cartilage. The protein is induced by transforming growth factor-beta and acts to inhibit cell adhesion. Mutations in this gene are associated with multiple types of corneal dystrophy.
Specifications

Specifications

TGFBI / BIGH3
0.39 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Beta ig h3, BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, Kerato epithelin, LCD1, RGD CAP, TGFBI
TGFBI
IgG2a
Caprylic Acid Precipitation
Antibody
3E11D11
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Mouse
P82198, Q15582
21810, 7045
TGFBI / BIGH3 Fusion Protein Ag0241
Primary
-20°C
TGFBI
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.