missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGFBI / BIGH3 Mouse anti-Human, Mouse, Clone: 3E11D11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60007-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes an RGD-containing protein that binds to type I, II and IV collagens. The RGD motif is found in many extracellular matrix proteins modulating cell adhesion and serves as a ligand recognition sequence for several integrins. This protein plays a role in cell-collagen interactions and may be involved in endochondrial bone formation in cartilage. The protein is induced by transforming growth factor-beta and acts to inhibit cell adhesion. Mutations in this gene are associated with multiple types of corneal dystrophy.Specifications
| TGFBI / BIGH3 | |
| Monoclonal | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P82198, Q15582 | |
| TGFBI | |
| TGFBI / BIGH3 Fusion Protein Ag0241 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 3E11D11 | |
| Unconjugated | |
| TGFBI | |
| Beta ig h3, BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, Kerato epithelin, LCD1, RGD CAP, TGFBI | |
| Mouse | |
| Caprylic Acid Precipitation | |
| RUO | |
| 21810, 7045 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction