Learn More
R&D Systems TGF-beta Premixed Magnetic Luminex™ Performance Assay
Click to view available options
Quantity:
2 Plex
3 Plex
Unit Size:
1 set
Specifications
Specifications
| Quantity | 3 Plex |
| Species | Human, Mouse, Rat, Porcine |
| Binding Target | TGF beta 1/2/3 |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7040 |
| Synonym | ARVD1, CED, DPD1, LAP, LDS5, RNHF, TGF beta1, TGFB, TGFB1, TGFB2, TGFB3, TGFbeta, TGFbeta 1, TGFbeta 2, TGFbeta 3, transforming growth factor, beta 1 |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |