Learn More
WTW™ TFK™ 325 Temperature Sensor
Product Code. 10697291
Change view
Click to view available options
Length (Metric) Cable:
1 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Cable unitSize
10697291 1 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10697291 Supplier WTW™ Supplier No. 102280
Only null left
Add to basket
Add to basket
Temperature sensors by WTW are used with pH and ORP electrodes that do not have their own temperature sensor
pH and ORP measurements must always have a temperature indication. During pH measurement, the indication of a temperature to compensate the temperature effect, and the documentation, with ORP measurement, only for documentation purposes. During ISE measurement it is important that the calibration and the measurement are performed in the same temperature range. Temperature sensors by WTW are used with pH and ORP electrodes that do not have their own temperature sensor. The temperature sensors are equipped with two 4 mm banana plugs.
TRUSTED_SUSTAINABILITY
Specifications
Type Temperature Sensor
Length (Metric) 1 m
Length (Metric) Cable 1 m
Temperature Sensor Yes