missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2,2-dimethyl-4H-pyran-4-one, 95%, Thermo Scientific Chemicals

Product Code. 15473777
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473777 250 mg 250mg
11362949 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473777 Supplier Thermo Scientific Alfa Aesar Supplier No. H27774.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1194-16-7 | Molecular Formula: C7H12O2 | Molecular Weight: 128.17 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1194-16-7
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD01549337
InChI Key BWMNOXJVRHGUQM-UHFFFAOYSA-N
Synonym 2,2-dimethyltetrahydropyran-4-one, 2,2-dimethyldihydro-2h-pyran-4 3h-one, 2,2-dimethyl-tetrahydro-pyran-4-one, tetrahydro-2,2-dimethyl-4h-pyran-4-one, 2,2-dimethyltetrahydro-4h-pyran-4-one, 4h-pyran-4-one, tetrahydro-2,2-dimethyl, 2,2-dimethyl-2h-3,5,6-trihydropyran-4-one, pubchem24423, acmc-1bt7l
PubChem CID 1738159
IUPAC Name 2,2-dimethyloxan-4-one
SMILES CC1(C)CC(=O)CCO1

Specifications

Boiling Point 60°C to 64°C (8 mmHg)
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 128.17
Percent Purity 95%
Chemical Name or Material Tetrahydro-2,2-dimethyl-4H-pyran-4-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.