accessibility menu, dialog, popup

UserName

CAS RN 1194-16-7

CH3H3COO

CAS RN 1194-16-7

IUPAC Name: 2,2-dimethyloxan-4-one
Synonyms: 2,2-dimethyloxan-4-one
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: BWMNOXJVRHGUQM-UHFFFAOYSA-N
SMILES: CC1(C)CC(=O)CCO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results