Learn More
WTW™ TetraCon™ Graphite 4-Electrode Conductivity Cell
Product Code. 10159271
Change view
Click to view available options
Material:
Graphite
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10159271 Graphite Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10159271 Supplier WTW™ Supplier No. 301602
Estimated Shipment: 07-12-2026
WTW carries two special measuring cells, which are specifically geared for these applications: The TetraCon™ S consists of an electrode body, which is perfectly fitted to measure pasteous samples thanks to its shovel shape and is easy to clean. The TetraCon™ 325/C is fitted with a chemical-resistant head made of PEEK to measure especially acidic samples.
TRUSTED_SUSTAINABILITY
Specifications
Description 4-electrode-conductivity cell
For Use With (Equipment) Special TetraCon™ 325/S Electrode System
For Use With (Application) Eliminate errors caused by polarization effects, and from dirty samples
Includes Built-in temperature sensor, 4.9 ft. cable, waterproof plug
Material Graphite
Temperature (Metric) Operating 0°C to 100°C
Length (Metric) Cable 1.5 m
Diameter (Metric) 15.3 mm