Learn More
WTW™ TetraCon™ 925-3 4 Electrode Conductivity Cell
GREENER_CHOICE
Product Code. 11725184
Click to view available options
Length (Metric) Cable:
3 m
Unit Size:
Each
Product Code. 11725184
Brand: WTW™ 301711
Only null left
Add to basket
Add to basket
With its easy handling and robust design, these electrode conductivity cells are proven 2 and 4 electrode technology.
  • IDS conductivity measuring cells – measurements from pure water all the way to concentrated solutions with proven two and four electrode technology
  • Proven 2 and 4 electrode technology
  • Easiest handling, robust design
  • Wide area of application - from pure water to highly concentrated solutions
TRUSTED_SUSTAINABILITY
Specifications
Description 4-electrodes, Graphite
Length (Metric) Cable 3 m
Temperature (Metric) Range 0°C to 100°C
Diameter (Metric) Body 15.3 mm
Length (Metric) 120 mm