Learn More
WTW™ TetraCon™ 325-10 Standard Conductivity Cell
Click to view available options
Length (Metric) Cable:
10 m
Unit Size:
Each
Specifications
Specifications
| Cell Constant | 0.475 cm-1 |
| For Use With (Application) | For conductivity measurements at depths down to 20m |
| Type | Standard Conductivity Cell |
| Diameter (Metric) Body | 15.3 mm |
| Length (Metric) Cable | 10 m |
| Material | Epoxy, Graphite |
| Includes | 4-electrode, temperature sensor, 10m Length cable and waterproof plug |
| Model | TetraCon™ 325-10 |