Learn More
- Home
- Products
- Baths
- Bath Accessories
- Memmert™ Test Tube Rack
Memmert™ Test Tube Rack
Product Code. 17512265 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
WTB35 Water Baths
WTB50 Water Baths
Holds:
100 Test Tubes (4 Rows)
30 Test Tubes (2 Rows)
44 Test Tubes (2 Rows)
50 Test Tubes (2 Rows)
58 Test Tubes (2 Rows)
66 Test Tubes (2 Rows)
74 Test Tubes (2 Rows)
75 Test Tubes (3 Rows)
Diameter (Metric):
10 mm
12 mm
14.5 mm
18 mm
21 mm
32 mm
Unit Size:
Each
| Product Code. | For Use With (Equipment) | Holds | Diameter (Metric) | unitSize |
|---|---|---|---|---|
| 17512265 | WTB35 Water Baths | 30 Test Tubes (2 Rows) | 32 mm | Each |
| 17502265 | WTB35 Water Baths | 44 Test Tubes (2 Rows) | 21 mm | Each |
| 17592255 | WTB35 Water Baths | 50 Test Tubes (2 Rows) | 18 mm | Each |
| 17582255 | WTB35 Water Baths | 58 Test Tubes (2 Rows) | 14.5 mm | Each |
| 17572255 | WTB35 Water Baths | 66 Test Tubes (2 Rows) | 12 mm | Each |
| 17562255 | WTB35 Water Baths | 74 Test Tubes (2 Rows) | 10 mm | Each |
| 17522265 | WTB35 Water Baths | 75 Test Tubes (3 Rows) | 18 mm | Each |
| 17532265 | WTB35 Water Baths | 100 Test Tubes (4 Rows) | 18 mm | Each |
| 17592265 | WTB50 Water Baths | 30 Test Tubes (2 Rows) | 32 mm | Each |
| 17582265 | WTB50 Water Baths | 44 Test Tubes (2 Rows) | 21 mm | Each |
| 17572265 | WTB50 Water Baths | 50 Test Tubes (2 Rows) | 18 mm | Each |
| 17562265 | WTB50 Water Baths | 58 Test Tubes (2 Rows) | 14.5 mm | Each |
| 17552265 | WTB50 Water Baths | 66 Test Tubes (2 Rows) | 12 mm | Each |
| 17542265 | WTB50 Water Baths | 74 Test Tubes (2 Rows) | 10 mm | Each |
| 17502275 | WTB50 Water Baths | 75 Test Tubes (3 Rows) | 18 mm | Each |
| 17512275 | WTB50 Water Baths | 100 Test Tubes (4 Rows) | 18 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Water Bath Accessory
Specifications
Specifications
| For Use With (Equipment) | WTB35 Water Baths |
| Type | Test Tube Rack |
| Holds | 30 Test Tubes (2 Rows) |
| Material | Stainless Steel |
| Diameter (Metric) | 32 mm |