Learn More
TES-1025, MedChemExpress
Product Code. 16483076
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16483076 25 mg 25mg
16495316 100 mg 100mg
16484428 10 mg 10mg
16430747 50 mg 50mg
16417157 5 mg 5mg
5 options
Product Code. 16483076
Brand: Medchem Express HY111365
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress TES-1025 is a potent and selective human α-amino-β-carboxymuconate-ε-semialdehyde decarboxylase (ACMSD) inhibitor with an IC50 of 13 nM.
TES-1025 is a potent and selective human α-amino-β-carboxymuconate-ε-semialdehyde decarboxylase (ACMSD) inhibitor with an IC50 of 13 nM.
Specifications
Chemical Name or Material TES-1025
CAS 1883602-21-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H13N3O3S2
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (260.80 mM; Need warming)
SMILES O=C1C(C#N)=C(C2=CC=CS2)N=C(SCC3=CC=CC(CC(O)=O)=C3)N1
Molecular Weight (g/mol) 383.44
Formula Weight 383.44
Percent Purity 95.11%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism