Learn More
TES-1025, MedChemExpress
Product Code. 16430747
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430747 50 mg 50mg
16495316 100 mg 100mg
16484428 10 mg 10mg
16483076 25 mg 25mg
16417157 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16430747 Supplier Medchem Express Supplier No. HY111365
MedChemExpress TES-1025 is a potent and selective human α-amino-β-carboxymuconate-ε-semialdehyde decarboxylase (ACMSD) inhibitor with an IC50 of 13 nM.
TES-1025 is a potent and selective human α-amino-β-carboxymuconate-ε-semialdehyde decarboxylase (ACMSD) inhibitor with an IC50 of 13 nM.
Specifications
Chemical Name or Material TES-1025
CAS 1883602-21-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H13N3O3S2
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (260.80 mM; Need warming)
SMILES O=C1C(C#N)=C(C2=CC=CS2)N=C(SCC3=CC=CC(CC(O)=O)=C3)N1
Molecular Weight (g/mol) 383.44
Formula Weight 383.44
Percent Purity 95.11%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism