Learn More
Dostmann Electronic™ Temperature Sensor, Thermocouple
Click to view available options
Length (Metric):
1000 mm
6000 mm
Temperature Range (Metric):
-40°C to +480°C
Up to 400°C
Unit Size:
Each
Description
- Type K according to EN 60584-1
- NiCr-Ni
- Mineral-insulated
Specifications
Specifications
| Diameter (Metric) Probe | 3 mm |
| Length (Metric) | 6000 mm |
| Product Type | Temperature Sensor |
| Description | Steel braiding with thermocouple strip |
| Temperature Range (Metric) | -40°C to +480°C |
| For Use With (Equipment) | P410 / TC301 / TC 309 / Mini-K |