Learn More
Testo™ Testoterm™ Temperature Strips
Product Code. 10028660
Change view
Click to view available options
Temperature Range (Metric):
37°C to 65°C
71°C to 110°C
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) unitSize
10028660 37°C to 65°C Pack of 10
10282161 71°C to 110°C Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10028660 Supplier Testo™ Supplier No. 0646 0108
  • ONLINE EXCLUSIVE PRICE
    438.71 NOK
    379.84 NOK / Pack of 10
    Save 58.87 NOK
    13% Off
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
Use these strips for monitoring the temperature of moving or small objects and long-term monitoring of transport and storage temperatures. Testoterm™ Temperature Strips can be applied to a variety of different surfaces and react to specific increases in temperature by changing color irreversibly within 2 to 3 seconds.
  • Self-adhesive temperature strips
  • Available in booklets of 10
  • Strips are peeled off and applied to the object you wish to measure
  • Change color when thresholds are exceeded within 2 to 3 seconds
Specifications
Mounting Self-adhesive
Accuracy (Temperature) ±1.5°C
Temperature Range (Metric) 37°C to 65°C
Type Self-adhesive Temperature Strips