Learn More
BINDER™ Calibration Certificate, Temperature and Humidity
This Temperature and Humidity Calibration Certificate is for use with BINDER™ KBWF 240 Growth Chambers with Light and Humidity.
Brand: BINDER™ 8012-1613
Additional Details : Weight : 0.00100kg
Description
- Calibration certificate for temperature and humidity measurement
- Calibrate to 25°C, 60% RH or at specified test values
- For use with BINDER™ KBWF 240 Growth Chambers with Light and Humidity
Specifications
BINDER Chambers KBWF 240 Growth Chambers with Light and Humidity | |
Calibration Certificate | |
BINDER™ KBWF | |
1 Year |
Operational Qualification | |
Measurement Including 27 Temperature Measuring Points and 1 Humidity Measuring Point at 25°C, 60% RH or (at Specified Values) | |
Temperature and Humidity | |
DIN 12880 |