Learn More
BINDER™ Temperature Calibration Certificate
Product Code. 30238264 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
KB PRO Series
KB Series
KBF PRO Series
KBF Series
Calibration Points:
15 to 18 Measuring Points
27 Measuring Points
28 Measuring Points
29 Measuring Points
30 Measuring Points
9 Measuring Points
Unit Size:
Each
Product Code. For Use With (Equipment) Calibration Points unitSize
30238264 KB PRO Series 28 Measuring Points Each
30238253 KB Series 9 Measuring Points Each
30238354 KB PRO Series 9 Measuring Points Each
30238322 KBF Series 9 Measuring Points Each
30238302 KBF PRO Series 9 Measuring Points Each
30238356 KB Series 15 to 18 Measuring Points Each
30238300 KB PRO Series 15 to 18 Measuring Points Each
30238231 KBF Series 15 to 18 Measuring Points Each
30238237 KBF PRO Series 15 to 18 Measuring Points Each
30238297 KB Series 27 Measuring Points Each
30238267 KBF Series 29 Measuring Points Each
30238215 KB PRO Series 30 Measuring Points Each
12 options
Product Code. 30238264
Brand: BINDER™ 80122425
Temperature measurement and certificate, measured at specified temperature.
  • Calibration measurement taken at specified temperature.
  • Choose between 9, 15 to 18, 27, 28, 29, or 30 measurement points
Specifications
For Use With (Equipment) KB PRO Series
Calibration Points 28 Measuring Points
Duration 1 Day
Product Type Calibration Certificate