Learn More
Fisherbrand™ Teclu Safety Burner with Tétine Connection, 13mm Opening
Shop All Fisherbrand Traceable ProductsDescription
- Safety features all contained within the burner
- For use with natural gas/methane or butane/propane
- 13mm diameter opening
- With gas control needle valve
- Specially designed conical mixing chamber incorporated which allows the burner to give off a very hot flame
- Suitable for glass work and soldering
- Gas and air mixing is regulated by adjusting the knurled wheel below the tube
Specifications
Specifications
| Product Type | Teclu Safety Burner |
| Gas Type | Butane, LPG, Propane |
| Diameter (Metric) | 13 mm |
| For Use With (Application) | Glasswork and soldering |
| Height (Metric) | 150 mm |
| Includes | T-Tine Connector |
| No. per Pack | 1 Ea. |