missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14606-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
May play an important role in sperm function and fertility.Specifications
TCP11 | |
Polyclonal | |
Unconjugated | |
TCP11 | |
D6S230E, KIAA0229, t complex 11 homolog (mouse), T complex protein 11 homolog, TCP11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21463, 309641, 6954 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q01755, Q5XI00, Q8WWU5 | |
TCP11 | |
TCP11 Fusion Protein Ag6160 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |