missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | TCP11 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883324
|
Proteintech
14606-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16873324
|
Proteintech
14606-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
May play an important role in sperm function and fertility.Specifications
TCP11 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q01755, Q5XI00, Q8WWU5 | |
21463, 309641, 6954 | |
TCP11 Fusion Protein Ag6160 | |
Primary | |
-20°C | |
TCP11 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D6S230E, KIAA0229, t complex 11 homolog (mouse), T complex protein 11 homolog, TCP11 | |
TCP11 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |