Learn More
Fisherbrand™ Tapered PTFE Stir Bars
Product Code. 16275761 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
10 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
45 mm
50 mm
55 mm
60 mm
70 mm
80 mm
Diameter (Metric):
4 mm
5 mm
7 mm
8 mm
10 mm
Unit Size:
Pack of 10
Pack of 5
Product Code. 16275761
Provides similar action as oval or pivot ring stirrers. Cowie Technology™ Tapered PTFE Stir Bars are stable and autoclavable for a wide variety of uses.
  • Stable, general-purpose magnetic stir bar
  • Tapered ends provide similar action as oval or pivot ring stirrers
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 7 mm
Length (Metric) 20 mm
Quantity 5/Pk.
Shape Tapered Cylindrical