Learn More
TAF12 Mouse anti-Human, Unconjugated, Clone: 4E12D2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67621-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16800356
  • 1611.84 NOK / 20µL
Estimated Shipment: 22-05-2024
Description
The TAF12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TAF12. This antibody recognizes human antigen. The TAF12 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TAF12
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
4E12D2
Unconjugated
20 μL
6883
IgG1