missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF12 Mouse anti-Human, Unconjugated, Clone: 4E12D2, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | TAF12 |
---|---|
Clone | 4E12D2 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16800356
|
Proteintech
67621-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16890346
|
Proteintech
67621-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The TAF12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TAF12. This antibody recognizes human antigen. The TAF12 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
TAF12 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
6883 | |
Primary |
4E12D2 | |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG1 |