Learn More
TAF12 Mouse anti-Human, Unconjugated, Clone: 4E12D2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67621-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The TAF12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TAF12. This antibody recognizes human antigen. The TAF12 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
TAF12 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
4E12D2 | |
Unconjugated | |
150 μL | |
6883 | |
IgG1 |