missing translation for 'onlineSavingsMsg'
Learn More

TACSTD2/TROP2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27360-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17679616

  • 4593.76 NOK / 150µL
Estimated Shipment: 03-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TACSTD2/TROP2 Polyclonal antibody apecifically detetects TACSTD2/TROP2 antigen in Human Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.
Specifications

Specifications

TACSTD2/TROP2
Polyclonal
Unconjugated
BC009409
Rabbit
Antigen affinity purification
Primary
Human
Liquid
Western Blot, Immunohistochemistry, ELISA
750 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
TACSTD2
TACSTD2/TROP2 fusion protein Ag25769
150 μL
4070
Store at -20°C. Stable for one year after shipment.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers